Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152721.5 | NP_689934.2 | 5, 6 | 800 | ||
XM_011525875.2 | XP_011524177.1 | 5, 6 | 821 | |||
XM_017025610.1 | XP_016881099.1 | 3, 4 | 2861 | |||
XM_017025611.1 | XP_016881100.1 | 3, 4 | 1375 | |||
GenBank mRNA | AK057795.1 | - | 4, 5 | 509 | ||
AY599248.1 | - | 5, 6 | 798 | |||
BC008583.1 | - | 3, 4 | 382 | |||
BC019045.2 | - | 3, 4 | 425 | |||
BC096744.1 | - | 5, 6 | 793 | 5260167 |