Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152742.2 | NP_689955.1 | 6 | 1231 | ||
GenBank mRNA | AK096281.1 | - | 6 | 1169 | ||
AL834418.1 | - | 6 | 1117 | |||
BC027972.1 | - | 6 | 1172 | 5240315 |