Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNPY4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152755.1 | NP_689968.1 | 1 | 189 | ||
GenBank mRNA | AK075537.1 | - | 1 | 143 | ||
AY359100.1 | - | 1 | 193 | |||
BC019903.1 | - | 1 | 119 | 4991480 | ||
BC032339.1 | - | 1 | 189 | 5191897 |