Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8orf46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152765.3 | NP_689978.2 | 5 | 821 | ||
GenBank mRNA | AK055047.1 | - | 2 | 155 | ||
AK096336.1 | - | 6 | 609 | |||
AK294261.1 | - | 4 | 409 | |||
AK308800.1 | - | 6 | 648 | |||
AK315486.1 | - | 5 | 564 | |||
BC028400.2 | - | 5 | 545 | 4816822 |