Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NANP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152667.2 | NP_689880.1 | 1, 2 | 162 | ||
GenBank mRNA | AK055472.1 | - | 1, 2 | 129 | ||
AL833116.1 | - | 1, 2 | 136 | |||
BC022552.1 | - | 1, 2 | 128 | 4819016 | ||
DA858639.1 | - | 1, 2 | 162 |