Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152696.3 | NP_689909.2 | 14 | 2958 | ||
NM_181358.2 | NP_852003.1 | 12 | 1678 | |||
NM_198268.2 | NP_938009.1 | 14 | 2958 | |||
NM_198269.2 | NP_938010.1 | 13 | 1869 | |||
XM_005270609.3 | XP_005270666.1 | 14 | 3135 | |||
XM_005270610.4 | XP_005270667.1 | 14 | 3132 | |||
XM_005270611.4 | XP_005270668.1 | 14 | 3135 | |||
XM_005270612.4 | XP_005270669.1 | 14 | 3097 | |||
XM_005270613.4 | XP_005270670.1 | 16 | 3048 | |||
XM_006710443.3 | XP_006710506.1 | 14 | 3033 | |||
XM_011540977.2 | XP_011539279.1 | 13 | 3015 | |||
XM_017000605.1 | XP_016856094.1 | 14 | 3094 | |||
GenBank mRNA | AB014530.1 | - | 5 | 638 | ||
AB089957.1 | - | 14 | 2922 | |||
AK122662.1 | - | 14 | 2956 | |||
AK302726.1 | - | 14 | 3142 | |||
AL833829.1 | - | 13 | 1847 | |||
BC028408.1 | - | 13 | 1874 | 4821915 | ||
BC033012.2 | - | 14 | 2958 | 4830408 | ||
BC036057.1 | - | 12 | 1678 | 5272816 |