Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130161.2 | NP_001123633.1 | 3 | 2471 | ||
NM_152709.4 | NP_689922.3 | 3 | 2471 | |||
XM_011539454.2 | XP_011537756.1 | 3 | 2202 | |||
GenBank mRNA | AK057864.1 | - | 1 | 90 | ||
AK057891.1 | - | 1 | 861 | |||
AK124681.1 | - | 3 | 2166 | |||
AK127878.1 | - | 1 | 1512 | |||
AY842014.1 | - | 3 | 2384 | |||
AY842017.1 | - | 3 | 2384 | |||
BC063627.1 | - | 1 | 852 |