Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5DC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152729.2 | NP_689942.2 | 10, 11 | 1186 | ||
GenBank mRNA | AK090918.1 | - | 7, 8 | 1916 | ||
AK290408.1 | - | 10, 11 | 1136 | |||
AK308337.1 | - | 10, 11 | 1169 | |||
AY203935.1 | - | 7, 8 | 797 | |||
BC015138.1 | - | 9, 10 | 966 | |||
BC108699.1 | - | 10, 11 | 1143 | 6061738 | ||
BC136832.1 | - | 10, 11 | 1145 | 9020823 | ||
BC136841.1 | - | 10, 11 | 1145 | 9020832 |