Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152743.3 | NP_689956.2 | 14 | 2899 | ||
XM_005249643.4 | XP_005249700.1 | 14 | 3041 | |||
XM_011515177.2 | XP_011513479.1 | 15 | 3130 | |||
XM_011515178.1 | XP_011513480.1 | 15 | 3341 | |||
XM_011515179.2 | XP_011513481.1 | 15 | 3121 | |||
XM_011515181.2 | XP_011513483.1 | 15 | 2941 | |||
XM_011515182.2 | XP_011513484.1 | 15 | 2916 | |||
XM_011515183.1 | XP_011513485.1 | 14 | 3010 | |||
XM_011515184.2 | XP_011513486.1 | 15 | 2916 | |||
XM_011515186.2 | XP_011513488.1 | 14 | 2769 | |||
XM_017011833.1 | XP_016867322.1 | 14 | 2842 | |||
XM_017011834.1 | XP_016867323.1 | 14 | 2853 | |||
XM_017011835.1 | XP_016867324.1 | 13 | 2746 | |||
XM_017011836.1 | XP_016867325.1 | 13 | 2682 | |||
GenBank mRNA | AK024482.1 | - | 11 | 4417 | ||
AK131083.1 | - | 11 | 5202 | |||
AK131097.1 | - | 3 | 5337 | |||
AL133088.1 | - | 7 | 1576 | |||
BC007209.1 | - | 10 | 2134 | |||
BC015632.2 | - | 14 | 2653 | 3839985 | ||
BC023561.2 | - | 10 | 1894 | |||
BC040704.1 | - | 1 | 1207 |