Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4orf22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206997.1 | NP_001193926.1 | 6 | 661 | ||
NM_152770.2 | NP_689983.2 | 5 | 610 | |||
XM_017007972.1 | XP_016863461.1 | 6 | 762 | |||
XM_017007975.1 | XP_016863464.1 | 4 | 750 | |||
XM_017007978.1 | XP_016863467.1 | 4 | 627 | |||
GenBank mRNA | AY349359.1 | - | 6 | 612 | ||
BC034296.1 | - | 5 | 610 | 5165558 | ||
DB522959.1 | - | 2 | 169 |