Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278629.1 | NP_001265558.1 | 3 | 649 | ||
NM_001278630.1 | NP_001265559.1 | 3 | 506 | |||
NM_152786.2 | NP_689999.1 | 3 | 722 | |||
XM_011518477.1 | XP_011516779.1 | 3 | 649 | |||
XM_011518478.1 | XP_011516780.1 | 3 | 649 | |||
GenBank mRNA | AK301872.1 | - | 3 | 506 | ||
AK313867.1 | - | 3 | 638 | |||
BC026884.1 | - | 3 | 688 | 3455891 | ||
BC033037.2 | - | 3 | 607 | 4827930 |