Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM133B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040057.2 | NP_001035146.1 | 3, 4 | 323 | ||
NM_001288584.1 | NP_001275513.1 | 2, 3 | 308 | |||
NM_152789.3 | NP_690002.2 | 2, 3 | 224 | |||
NR_109929.1 | 2, 3 | 224 | ||||
XM_011516004.2 | XP_011514306.1 | 2, 3 | 487 | |||
XM_011516005.2 | XP_011514307.1 | 2, 3 | 224 | |||
GenBank mRNA | AK125498.1 | - | 2, 3 | 220 | ||
AK313692.1 | - | 2, 3 | 308 | |||
AL833918.2 | - | 3, 4 | 323 | |||
BC017874.1 | - | 1 | 220 | |||
BC032461.1 | - | 2, 3 | 169 | 5197918 | ||
BC057771.1 | - | 3, 4 | 314 | 5301001 | ||
BC062706.1 | - | 2, 3 | 228 | |||
BC090868.1 | - | 2, 3 | 207 | 6190536 | ||
BX337761.2 | - | 2, 3 | 159 |
Gene Symbol: FAM133CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027508.1 | 1 | 660 |
Gene Symbol: FAM133DP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034169.1 | 1 | 216 |
Gene Symbol: LOC100421561 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC157872.1 | - | 1 | 181 | 9057091 |