Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TANGO2 SIRNA Assays ›
Gene Symbol:
TANGO2
Gene Name:
transport and golgi organization 2 homolog
Gene Aliases:
C22orf25, MECRCN
Chromosome Location:
Chr.22: 20017000 - 20067164 on Build GRCh38
UniGene ID:
Hs.474233
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
39734
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TANGO2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001283106.2
NP_001270035.1
4
436
NM_001283116.2
NP_001270045.1
3
190
NM_001283129.2
NP_001270058.1
3
430
NM_001283148.2
NP_001270077.1
3
190
NM_001283154.2
NP_001270083.1
3
302
NM_001283179.2
NP_001270108.1
3
317
NM_001283186.2
NP_001270115.1
3
190
NM_001283199.2
NP_001270128.1
3
302
NM_001283215.2
NP_001270144.1
3
430
NM_001283235.2
NP_001270164.1
3
190
NM_001283248.2
NP_001270177.1
3
190
NM_001322141.1
NP_001309070.1
3
430
NM_001322142.1
NP_001309071.1
3
190
NM_001322143.1
NP_001309072.1
3
503
NM_001322144.1
NP_001309073.1
3
430
NM_001322145.1
NP_001309074.1
3
503
NM_001322146.1
NP_001309075.1
3
263
NM_001322147.1
NP_001309076.1
3
430
NM_001322148.1
NP_001309077.1
3
430
NM_001322149.1
NP_001309078.1
3
430
NM_001322150.1
NP_001309079.1
3
430
NM_001322153.1
NP_001309082.1
3
190
NM_001322155.1
NP_001309084.1
3
263
NM_001322160.1
NP_001309089.1
3
315
NM_001322163.1
NP_001309092.1
4
436
NM_001322166.1
NP_001309095.1
3
263
NM_001322167.1
NP_001309096.1
3
315
NM_001322169.1
NP_001309098.1
3
190
NM_001322171.1
NP_001309100.1
3
263
NM_001322172.1
NP_001309101.1
3
555
NM_001322173.1
NP_001309102.1
3
430
NM_001322174.1
NP_001309103.1
3
317
NM_001322175.1
NP_001309104.1
3
315
NM_152906.6
NP_690870.3
3
302
NR_104274.2
3
190
NR_104275.2
3
430
NR_136206.1
3
263
NR_136211.1
3
263
NR_136212.1
3
263
XM_011529863.1
XP_011528165.1
3
264
XM_011529865.1
XP_011528167.1
2
206
XM_011529867.1
XP_011528169.1
4
400
XM_017028577.1
XP_016884066.1
3
263
XM_017028578.1
XP_016884067.1
3
282
XM_017028579.1
XP_016884068.1
3
311
XM_017028580.1
XP_016884069.1
4
400
XM_017028581.1
XP_016884070.1
3
317
XM_017028582.1
XP_016884071.1
4
400
XM_017028583.1
XP_016884072.1
3
147
XM_017028584.1
XP_016884073.1
3
410
XM_017028585.1
XP_016884074.1
3
262
XM_017028586.1
XP_016884075.1
3
187
XM_017028587.1
XP_016884076.1
3
317
XM_017028588.1
XP_016884077.1
3
243
GenBank mRNA
AK092484.1
-
3
145
AK289862.1
-
4
405
AK295210.1
-
2
316
AK297075.1
-
3
147
AK298593.1
-
3
148
AK301054.1
-
3
149
AK301366.1
-
3
389
AK309398.1
-
2
70
AK311676.1
-
3
149
AK316020.1
-
3
151
AK316056.1
-
3
410
BC041339.1
-
3
265
5284126
BG685068.1
-
1
40
BI544307.1
-
3
265
BI911898.1
-
3
253
BP193987.1
-
3
388
BP303570.1
-
2
87
BU173566.1
-
3
222
BX345246.2
-
3
302
CD675132.1
-
3
244
CF552364.1
-
2
161
CR456355.1
-
2
117
CR627452.1
-
3
145
DA176876.1
-
3
188
DC303300.1
-
4
405
DC315148.1
-
3
148
DC318353.1
-
3
190
DC332363.1
-
3
410
DC349456.1
-
3
386
HY135099.1
-
3
317
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-x88h5:80/100.66.78.96:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline