Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152945.3 | NP_694453.2 | 1, 2 | 431 | ||
XM_005246287.4 | XP_005246344.1 | 1, 2 | 441 | |||
XM_017003322.1 | XP_016858811.1 | 1, 2 | 440 | |||
GenBank mRNA | AB036991.1 | - | 1, 2 | 353 | ||
AF526533.1 | - | 1, 2 | 431 | |||
AK126575.1 | - | 1, 2 | 289 | |||
BC066549.1 | - | 1, 2 | 400 | 5748065 | ||
BI763677.1 | - | 1, 2 | 431 |