Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ODF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153007.4 | NP_694552.2 | 1 | 615 | ||
XM_011523688.1 | XP_011521990.1 | 1 | 67 | |||
GenBank mRNA | AB081120.1 | - | 1 | 615 | ||
AY237799.1 | - | 1 | 615 | |||
BC112010.1 | - | 1 | 529 | 8327478 | ||
BC112014.1 | - | 1 | 529 | 8327482 |