Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001702.2 | NP_001693.2 | 1 | 335 | ||
XM_011517199.2 | XP_011515501.1 | 2 | 773 | |||
XM_011517202.2 | XP_011515504.1 | 2 | 773 | |||
XM_017013691.1 | XP_016869180.1 | 2 | 773 | |||
XM_017013692.1 | XP_016869181.1 | 2 | 773 | |||
XM_017013693.1 | XP_016869182.1 | 2 | 773 | |||
XM_017013694.1 | XP_016869183.1 | 2 | 773 | |||
XM_017013695.1 | XP_016869184.1 | 2 | 773 | |||
XM_017013696.1 | XP_016869185.1 | 2 | 773 | |||
XM_017013697.1 | XP_016869186.1 | 2 | 773 | |||
GenBank mRNA | AB005297.1 | - | 1 | 335 |
Set Membership: |
G-protein coupled receptor |