Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM218A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153027.1 | NP_694572.1 | 1 | 373 | ||
GenBank mRNA | AK056221.1 | - | 1 | 373 | ||
BC022381.1 | - | 1 | 341 | 4248805 |