Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153213.3 | NP_694945.2 | 2 | 232 | ||
XM_011540706.2 | XP_011539008.1 | 3 | 396 | |||
GenBank mRNA | AK091281.1 | - | 2 | 268 | ||
AK297707.1 | - | 2 | 294 | |||
BC040640.1 | - | 2 | 232 | 3847806 |