Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039547.2 | NP_001034636.1 | 4 | 485 | ||
NR_033289.1 | 4 | 485 | ||||
XM_011512624.2 | XP_011510926.1 | 4 | 283 | |||
XM_017006088.1 | XP_016861577.1 | 4 | 470 | |||
XM_017006089.1 | XP_016861578.1 | 4 | 453 | |||
XM_017006090.1 | XP_016861579.1 | 4 | 470 | |||
GenBank mRNA | AK127641.1 | - | 4 | 453 | ||
AK310761.1 | - | 4 | 470 | |||
AK313792.1 | - | 4 | 367 | |||
BC032470.1 | - | 4 | 445 | 5217372 | ||
BX648359.1 | - | 4 | 465 | |||
BX648681.1 | - | 4 | 426 | |||
DR000158.1 | - | 4 | 486 |