Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152778.2 | NP_689991.1 | 6 | 660 | ||
XM_005262893.1 | XP_005262950.1 | 5 | 574 | |||
XM_005262896.1 | XP_005262953.1 | 5 | 574 | |||
XM_005262897.2 | XP_005262954.1 | 5 | 574 | |||
XM_005262898.2 | XP_005262955.1 | 5 | 574 | |||
XM_017007989.1 | XP_016863478.1 | 5 | 574 | |||
GenBank mRNA | AK074564.1 | - | 5 | 509 | ||
AK294527.1 | - | 5 | 483 | |||
AK294957.1 | - | 5 | 502 | |||
AK315596.1 | - | 5 | 506 | |||
BC029503.1 | - | 6 | 625 | 5272664 | ||
BI553701.1 | - | 5 | 574 |