Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CATSPERD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152784.3 | NP_689997.3 | 19 | 1746 | ||
XM_011527885.2 | XP_011526187.1 | 19 | 2038 | |||
XM_011527886.2 | XP_011526188.1 | 19 | 1984 | |||
XM_011527888.2 | XP_011526190.1 | 19 | 2038 | |||
XM_011527889.2 | XP_011526191.1 | 19 | 2038 | |||
XM_011527891.1 | XP_011526193.1 | 10 | 834 | |||
XM_011527892.1 | XP_011526194.1 | 8 | 555 | |||
XM_017026564.1 | XP_016882053.1 | 18 | 1954 | |||
XM_017026565.1 | XP_016882054.1 | 18 | 1691 | |||
XM_017026566.1 | XP_016882055.1 | 15 | 1624 | |||
GenBank mRNA | AK128088.1 | - | 19 | 1754 | ||
BC031233.1 | - | 8 | 696 | |||
BC043005.1 | - | 19 | 1732 | 5297674 |