Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153217.2 | NP_694949.1 | 1 | 246 | ||
GenBank mRNA | AK055830.1 | - | 1 | 228 | ||
AK315460.1 | - | 1 | 246 | |||
BC019346.1 | - | 2 | 255 | 3613225 |