Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKAIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152864.3 | NP_690603.3 | 7 | 1398 | ||
XM_005260192.1 | XP_005260249.1 | 6 | 1275 | |||
XM_005260194.3 | XP_005260251.1 | 6 | 1734 | |||
XM_011528527.1 | XP_011526829.1 | 7 | 1606 | |||
XM_011528528.2 | XP_011526830.1 | 7 | 2068 | |||
XM_011528529.2 | XP_011526831.1 | 7 | 2054 | |||
XM_017027636.1 | XP_016883125.1 | 7 | 1576 | |||
XM_017027637.1 | XP_016883126.1 | 7 | 2058 | |||
XM_017027638.1 | XP_016883127.1 | 7 | 1818 | |||
XM_017027639.1 | XP_016883128.1 | 7 | 1804 | |||
GenBank mRNA | AK126246.1 | - | 7 | 1321 | ||
BC041812.1 | - | 7 | 1396 | 5302801 |