Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGKD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003648.2 | NP_003639.2 | 27 | 3330 | ||
NM_152879.2 | NP_690618.2 | 28 | 3395 | |||
XM_005246107.3 | XP_005246164.1 | 26 | 3062 | |||
XM_005246108.1 | XP_005246165.1 | 26 | 3216 | |||
XM_005246110.3 | XP_005246167.1 | 17 | 3067 | |||
XM_011512028.2 | XP_011510330.1 | 28 | 4854 | |||
XM_011512029.2 | XP_011510331.1 | 28 | 4855 | |||
XM_011512030.2 | XP_011510332.1 | 28 | 3451 | |||
XM_011512031.2 | XP_011510333.1 | 28 | 3325 | |||
XM_011512032.2 | XP_011510334.1 | 28 | 3309 | |||
XM_011512035.1 | XP_011510337.1 | 26 | 3196 | |||
XM_011512037.2 | XP_011510339.1 | 26 | 3300 | |||
XM_011512040.2 | XP_011510342.1 | 26 | 3232 | |||
XM_017005138.1 | XP_016860627.1 | 28 | 3733 | |||
XM_017005139.1 | XP_016860628.1 | 26 | 3225 | |||
XM_017005140.1 | XP_016860629.1 | 28 | 3327 | |||
GenBank mRNA | AB078966.1 | - | 28 | 3395 | ||
BC006561.2 | - | 2 | 184 | |||
D63479.3 | - | 28 | 3336 | |||
D73409.1 | - | 27 | 3328 |