Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ODF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319953.1 | NP_001306882.1 | 3 | 634 | ||
NM_153007.4 | NP_694552.2 | 3 | 949 | |||
XM_005256476.3 | XP_005256533.1 | 4 | 802 | |||
XM_011523688.1 | XP_011521990.1 | 4 | 571 | |||
GenBank mRNA | AB081120.1 | - | 3 | 949 | ||
AY237799.1 | - | 3 | 949 | |||
BC112010.1 | - | 3 | 863 | 8327478 | ||
BC112014.1 | - | 3 | 863 | 8327482 | ||
FJ235526.1 | - | 3 | 604 |