Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305121.1 | NP_001292050.1 | 9 | 1558 | ||
NM_001305122.1 | NP_001292051.1 | 9 | 1538 | |||
NM_153208.2 | NP_694940.1 | 10 | 1623 | |||
NR_130967.1 | 8 | 1492 | ||||
NR_130968.1 | 9 | 1525 | ||||
XM_011545736.2 | XP_011544038.1 | 9 | 958 | |||
XM_017022933.1 | XP_016878422.1 | 9 | 1669 | |||
GenBank mRNA | AF520569.1 | - | 8 | 1492 | ||
AF520570.1 | - | 10 | 1623 | |||
AK093894.1 | - | 7 | 763 | |||
AK301933.1 | - | 8 | 861 | |||
AK303998.1 | - | 8 | 851 | |||
AK311216.1 | - | 8 | 818 | |||
BC010515.1 | - | 4 | 434 | |||
BC034823.1 | - | 9 | 954 | 5165639 |