Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP120 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166226.1 | NP_001159698.1 | 10, 11 | 1623 | ||
NM_153223.3 | NP_694955.2 | 11, 12 | 1704 | |||
XM_005271901.4 | XP_005271958.1 | 10, 11 | 1485 | |||
XM_011543185.2 | XP_011541487.1 | 10, 11 | 2038 | |||
XM_011543186.2 | XP_011541488.1 | 7, 8 | 1317 | |||
XM_017009085.1 | XP_016864574.1 | 7, 8 | 1182 | |||
GenBank mRNA | AK093409.1 | - | 3, 4 | 390 | ||
AK095646.1 | - | 10, 11 | 1625 | |||
AK296999.1 | - | 11, 12 | 1668 | |||
BC040527.1 | - | 10, 11 | 2287 | |||
BC144558.1 | - | 13, 14 | 1831 | |||
BX648687.1 | - | 1, 2 | 97 |