Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286434.1 | NP_001273363.1 | 2 | 212 | ||
NM_153356.2 | NP_699187.1 | 3 | 320 | |||
XM_006720409.3 | XP_006720472.1 | 3 | 350 | |||
XM_006720410.3 | XP_006720473.1 | 2 | 242 | |||
XM_011521281.2 | XP_011519583.1 | 3 | 349 | |||
XM_011521282.2 | XP_011519584.1 | 3 | 349 | |||
XM_011521283.2 | XP_011519585.1 | 3 | 349 | |||
XM_011521284.2 | XP_011519586.1 | 2 | 129 | |||
GenBank mRNA | AB449907.1 | - | 2 | 88 | ||
BC033516.1 | - | 3 | 279 | 5170475 |