Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LACC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128303.1 | NP_001121775.1 | 2 | 654 | ||
NM_153218.2 | NP_694950.2 | 2 | 490 | |||
XM_005266261.3 | XP_005266318.1 | 2 | 1113 | |||
XM_006719766.3 | XP_006719829.1 | 2 | 271 | |||
XM_006719767.3 | XP_006719830.1 | 2 | 490 | |||
XM_011534933.2 | XP_011533235.1 | 3 | 1308 | |||
XM_011534934.2 | XP_011533236.1 | 2 | 427 | |||
XM_011534935.2 | XP_011533237.1 | 2 | 490 | |||
XM_017020394.1 | XP_016875883.1 | 2 | 547 | |||
GenBank mRNA | AK096044.1 | - | 2 | 490 | ||
BC035749.1 | - | 2 | 459 | 5750179 | ||
BX503941.1 | - | 2 | 381 | |||
DR001026.1 | - | 2 | 381 |