Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP120 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166226.1 | NP_001159698.1 | 11, 12 | 1805 | ||
NM_153223.3 | NP_694955.2 | 12, 13 | 1886 | |||
XM_005271901.4 | XP_005271958.1 | 11, 12 | 1667 | |||
XM_011543185.2 | XP_011541487.1 | 11, 12 | 2220 | |||
XM_011543186.2 | XP_011541488.1 | 8, 9 | 1499 | |||
XM_017009085.1 | XP_016864574.1 | 8, 9 | 1364 | |||
GenBank mRNA | AK093409.1 | - | 4, 5 | 572 | ||
AK095646.1 | - | 11, 12 | 1807 | |||
AK296999.1 | - | 12, 13 | 1850 | |||
BC040527.1 | - | 11, 12 | 2469 | |||
BC144558.1 | - | 14, 15 | 2013 | |||
BX648687.1 | - | 2, 3 | 279 |