Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRWD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153252.4 | NP_694984.4 | 20 | 2531 | ||
XM_005262113.3 | XP_005262170.1 | 20 | 2533 | |||
XM_017029384.1 | XP_016884873.1 | 16 | 2061 | |||
XM_017029385.1 | XP_016884874.1 | 20 | 2533 | |||
GenBank mRNA | AK095887.1 | - | 3 | 404 | ||
AY497046.1 | - | 20 | 2531 | |||
AY497047.1 | - | 17 | 2463 | |||
AY497048.1 | - | 20 | 2554 | |||
AY497049.1 | - | 19 | 2524 | |||
AY497050.1 | - | 20 | 2504 | |||
AY497051.1 | - | 17 | 2091 | |||
AY497052.1 | - | 22 | 2690 | |||
AY497053.1 | - | 18 | 2279 | |||
AY497054.1 | - | 17 | 2180 | |||
AY497055.1 | - | 20 | 2465 | |||
AY497056.1 | - | 17 | 2436 | |||
AY497057.1 | - | 16 | 2335 | |||
AY497058.1 | - | 16 | 2174 | |||
AY497059.1 | - | 18 | 2545 | |||
AY497060.1 | - | 14 | 2112 | |||
BC111490.1 | - | 20 | 2481 |