Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNEP1R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281789.1 | NP_001268718.1 | 6 | 597 | ||
NM_153261.5 | NP_694993.2 | 7 | 648 | |||
NR_104042.1 | 6 | 574 | ||||
NR_104043.1 | 5 | 523 | ||||
GenBank mRNA | AK095420.1 | - | 6 | 570 | ||
AK301610.1 | - | 7 | 613 | |||
AK307132.1 | - | 6 | 644 | |||
BC022550.1 | - | 5 | 474 | |||
BC036683.2 | - | 7 | 577 | 5266084 | ||
CR933600.1 | - | 7 | 593 | |||
FJ516381.1 | - | 6 | 525 |