Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCEAL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006684.1 | NP_001006685.1 | 2 | 252 | ||
NM_153333.2 | NP_699164.1 | 3 | 325 | |||
GenBank mRNA | AK026349.1 | - | 3 | 302 | ||
BC035573.1 | - | 3 | 342 | 5405281 | ||
BU932815.1 | - | 3 | 337 |