Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKAIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040214.2 | NP_001035304.1 | 2, 3 | 553 | ||
NM_001300737.1 | NP_001287666.1 | 3, 4 | 617 | |||
NM_001300738.1 | NP_001287667.1 | 2, 3 | 553 | |||
NM_001300740.1 | NP_001287669.1 | 2, 3 | 191 | |||
NM_153355.4 | NP_699186.2 | 2, 3 | 553 | |||
XM_011535501.2 | XP_011533803.1 | 2, 3 | 441 | |||
XM_011535502.2 | XP_011533804.1 | 2, 3 | 208 | |||
XM_017010319.1 | XP_016865808.1 | 2, 3 | 441 | |||
GenBank mRNA | AB070452.1 | - | 2, 3 | 362 | ||
AK055326.1 | - | 2, 3 | 186 | |||
AK308927.1 | - | 2, 3 | 191 | |||
AK309802.1 | - | 2, 3 | 191 | |||
AK310763.1 | - | 2, 3 | 228 | |||
AK315886.1 | - | 2, 3 | 192 | |||
BC035062.2 | - | 3, 4 | 319 | 5264304 | ||
DB174077.1 | - | 1, 2 | 228 |