Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153365.2 | NP_699196.2 | 2 | 330 | ||
XM_005248139.2 | XP_005248196.1 | 2 | 263 | |||
XM_005248140.3 | XP_005248197.1 | 2 | 213 | |||
XM_011513812.2 | XP_011512114.1 | 2 | 328 | |||
XM_011513815.2 | XP_011512117.1 | 2 | 1548 | |||
XM_017007876.1 | XP_016863365.1 | 2 | 154 | |||
GenBank mRNA | AK074494.1 | - | 2 | 263 | ||
BC066899.1 | - | 2 | 268 | 4825556 | ||
DB461077.1 | - | 2 | 333 |