Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRIG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136051.2 | NP_001129523.1 | 13 | 1429 | ||
NM_153377.4 | NP_700356.2 | 13 | 1848 | |||
XM_011537893.2 | XP_011536195.1 | 13 | 1520 | |||
XM_017018790.1 | XP_016874279.1 | 13 | 1850 | |||
GenBank mRNA | AK074921.1 | - | 4 | 348 | ||
AK292857.1 | - | 13 | 1829 | |||
AK301162.1 | - | 13 | 1409 | |||
AL833710.1 | - | 2 | 185 | |||
AY358288.1 | - | 13 | 1791 | |||
AY358295.1 | - | 13 | 1429 | |||
AY505340.1 | - | 13 | 1599 | |||
BC126169.1 | - | 13 | 1744 | 8991885 | ||
BC126171.1 | - | 13 | 1744 | 8991887 | ||
BC143960.1 | - | 13 | 1744 |