Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP120 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166226.1 | NP_001159698.1 | 14, 15 | 2146 | ||
NM_153223.3 | NP_694955.2 | 15, 16 | 2227 | |||
XM_005271901.4 | XP_005271958.1 | 14, 15 | 2008 | |||
XM_011543185.2 | XP_011541487.1 | 14, 15 | 2561 | |||
XM_011543186.2 | XP_011541488.1 | 11, 12 | 1840 | |||
XM_017009085.1 | XP_016864574.1 | 11, 12 | 1705 | |||
GenBank mRNA | AK093409.1 | - | 7, 8 | 913 | ||
AK095646.1 | - | 14, 15 | 2148 | |||
AK296999.1 | - | 13, 14 | 1921 | |||
AL833929.1 | - | 3, 4 | 334 | |||
BC040527.1 | - | 14, 15 | 2810 | |||
BC144558.1 | - | 17, 18 | 2354 | |||
BX648687.1 | - | 5, 6 | 620 |