Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF550 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277090.1 | NP_001264019.1 | 6 | 2988 | ||
NM_001277091.1 | NP_001264020.1 | 6 | 2402 | |||
NM_001277092.1 | NP_001264021.1 | 5 | 2055 | |||
NM_001277093.1 | NP_001264022.1 | 5 | 680 | |||
XM_011526567.2 | XP_011524869.1 | 5 | 3392 | |||
XM_011526568.2 | XP_011524870.1 | 5 | 4485 | |||
XM_017026401.1 | XP_016881890.1 | 6 | 2333 | |||
GenBank mRNA | AK122867.1 | - | 5 | 2056 | ||
AL833214.1 | - | 2 | 2964 | |||
BC034810.1 | - | 6 | 2378 | 4823435 | ||
BC053858.1 | - | 6 | 2962 | 6044705 | ||
BX538032.1 | - | 5 | 2042 | |||
CX756948.1 | - | 5 | 641 |