Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRWD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153252.4 | NP_694984.4 | 33, 34 | 4078 | ||
XM_005262113.3 | XP_005262170.1 | 32, 33 | 3930 | |||
XM_017029384.1 | XP_016884873.1 | 29, 30 | 3608 | |||
GenBank mRNA | AK095887.1 | - | 16, 17 | 1951 | ||
AY497046.1 | - | 33, 34 | 4078 | |||
AY497047.1 | - | 30, 31 | 4010 | |||
AY497048.1 | - | 33, 34 | 4101 | |||
AY497049.1 | - | 32, 33 | 4071 | |||
AY497050.1 | - | 33, 34 | 4051 | |||
AY497051.1 | - | 30, 31 | 3638 | |||
AY497052.1 | - | 35, 36 | 4237 | |||
AY497053.1 | - | 31, 32 | 3826 | |||
AY497054.1 | - | 30, 31 | 3727 | |||
AY497055.1 | - | 33, 34 | 4012 | |||
AY497056.1 | - | 30, 31 | 3983 | |||
AY497057.1 | - | 29, 30 | 3882 | |||
AY497058.1 | - | 29, 30 | 3721 | |||
AY497059.1 | - | 31, 32 | 4092 | |||
AY497060.1 | - | 27, 28 | 3659 | |||
BC039508.2 | - | 5, 6 | 551 | |||
CB269573.1 | - | 4, 5 | 452 |