Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATXN7L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153340.4 | NP_699171.3 | 10 | 1800 | ||
XM_005270445.3 | XP_005270502.1 | 10 | 1931 | |||
XM_005270446.2 | XP_005270503.1 | 10 | 1927 | |||
XM_011540638.2 | XP_011538940.1 | 9 | 1830 | |||
XM_011540639.2 | XP_011538941.1 | 10 | 3200 | |||
XM_011540641.2 | XP_011538943.1 | 10 | 2533 | |||
XM_011540644.2 | XP_011538946.1 | 12 | 2015 | |||
XM_011540645.1 | XP_011538947.1 | 9 | 1776 | |||
XM_017000267.1 | XP_016855756.1 | 11 | 2662 | |||
XM_017000268.1 | XP_016855757.1 | 11 | 1995 | |||
XM_017000269.1 | XP_016855758.1 | 11 | 2662 | |||
XM_017000270.1 | XP_016855759.1 | 11 | 2063 | |||
GenBank mRNA | AK090460.1 | - | 7 | 3478 |