Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKAIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040214.2 | NP_001035304.1 | 7 | 2667 | ||
NM_001300737.1 | NP_001287666.1 | 8 | 2731 | |||
NM_001300738.1 | NP_001287667.1 | 8 | 2681 | |||
NM_001300740.1 | NP_001287669.1 | 7 | 2305 | |||
NM_153355.4 | NP_699186.2 | 6 | 2466 | |||
XM_011535501.2 | XP_011533803.1 | 8 | 2644 | |||
XM_011535502.2 | XP_011533804.1 | 8 | 2411 | |||
XM_011535503.2 | XP_011533805.1 | 7 | 2681 | |||
XM_017010318.1 | XP_016865807.1 | 6 | 2590 | |||
XM_017010319.1 | XP_016865808.1 | 7 | 2443 | |||
GenBank mRNA | AK055326.1 | - | 7 | 2299 | ||
BC035062.2 | - | 8 | 2433 | 5264304 |