Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPNE8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153634.2 | NP_705898.1 | 6 | 473 | ||
XM_011537951.2 | XP_011536253.1 | 6 | 3448 | |||
XM_011537952.2 | XP_011536254.1 | 6 | 3448 | |||
XM_017018851.1 | XP_016874340.1 | 7 | 1446 | |||
XM_017018852.1 | XP_016874341.1 | 6 | 419 | |||
GenBank mRNA | AK098593.1 | - | 6 | 403 | ||
AK307908.1 | - | 6 | 446 | |||
AY177785.1 | - | 6 | 389 | |||
BC068040.1 | - | 6 | 437 | |||
BC110577.1 | - | 6 | 464 | 40007743 | ||
BC110578.1 | - | 6 | 464 | 40007744 | ||
BI669075.1 | - | 6 | 446 | |||
BX538055.1 | - | 6 | 487 |