Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPNE9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308388.1 | NP_001295317.1 | 4 | 369 | ||
NM_153635.2 | NP_705899.2 | 4 | 350 | |||
XM_011533386.2 | XP_011531688.1 | 3 | 526 | |||
XM_011533388.2 | XP_011531690.1 | 3 | 526 | |||
XM_011533389.2 | XP_011531691.1 | 3 | 526 | |||
XM_017005747.1 | XP_016861236.1 | 4 | 172 | |||
GenBank mRNA | BC035735.1 | - | 4 | 350 | 5729888 | |
BC130375.1 | - | 4 | 221 | 40146406 | ||
BM545574.1 | - | 4 | 350 | |||
HY151604.1 | - | 4 | 370 |