Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320480.1 | NP_001307409.1 | 5 | 659 | ||
NM_016430.3 | NP_057514.2 | 6 | 673 | |||
NM_153681.2 | NP_710148.1 | 4 | 614 | |||
NM_153682.2 | NP_710149.1 | 5 | 507 | |||
XM_005260990.4 | XP_005261047.1 | 5 | 372 | |||
XM_011529596.2 | XP_011527898.1 | 5 | 406 | |||
XM_017028364.1 | XP_016883853.1 | 5 | 424 | |||
XM_017028365.1 | XP_016883854.1 | 6 | 615 | |||
GenBank mRNA | AB035742.1 | - | 5 | 439 | ||
AB035743.1 | - | 5 | 369 | |||
AB035744.1 | - | 5 | 401 | |||
AB035745.1 | - | 4 | 421 | |||
AB037162.1 | - | 5 | 443 | |||
AB037163.1 | - | 4 | 412 | |||
AB039659.1 | - | 4 | 326 | |||
AB451328.1 | - | 3 | 248 | |||
AB451472.1 | - | 3 | 248 | |||
AF237812.1 | - | 4 | 463 | |||
AK026311.1 | - | 4 | 399 | |||
AK314457.1 | - | 4 | 407 | |||
AK316609.1 | - | 4 | 344 | |||
BC005180.1 | - | 5 | 388 | 3929930 | ||
BC011007.1 | - | 5 | 392 | 4281552 | ||
BE275469.1 | - | 5 | 506 | |||
BT007053.1 | - | 4 | 326 |