Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318469.1 | NP_001305398.1 | 4 | 513 | ||
NM_001318471.1 | NP_001305400.1 | 5 | 569 | |||
NM_001318472.1 | NP_001305401.1 | 3 | 436 | |||
NM_001318473.1 | NP_001305402.1 | 3 | 386 | |||
NM_001318474.1 | NP_001305403.1 | 3 | 367 | |||
NM_001318475.1 | NP_001305404.1 | 4 | 525 | |||
NM_001318476.1 | NP_001305405.1 | 4 | 511 | |||
NM_153688.3 | NP_710155.2 | 4 | 494 | |||
XM_011522921.2 | XP_011521223.1 | 6 | 554 | |||
XM_017022983.1 | XP_016878472.1 | 4 | 681 | |||
XM_017022984.1 | XP_016878473.1 | 5 | 808 | |||
XM_017022985.1 | XP_016878474.1 | 5 | 813 | |||
XM_017022986.1 | XP_016878475.1 | 6 | 901 | |||
XM_017022987.1 | XP_016878476.1 | 4 | 525 | |||
GenBank mRNA | AK094761.1 | - | 4 | 431 | ||
AK291372.1 | - | 3 | 314 | |||
AK296560.1 | - | 4 | 443 | |||
AK308134.1 | - | 3 | 343 | |||
AL038510.1 | - | 4 | 412 | |||
AL538214.3 | - | 4 | 381 | |||
BC064604.1 | - | 4 | 372 | |||
BC113863.1 | - | 2 | 185 | |||
BC114471.1 | - | 2 | 185 | |||
BC127184.1 | - | 2 | 185 | |||
BX647144.1 | - | 2 | 149 | |||
DA781502.1 | - | 3 | 290 | |||
DC371394.1 | - | 3 | 314 | |||
DC388176.1 | - | 4 | 451 | |||
DC402593.1 | - | 5 | 516 |