Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDELC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153705.4 | NP_714916.3 | 2, 3 | 474 | ||
XM_011542621.2 | XP_011540923.1 | 2, 3 | 447 | |||
XM_011542623.2 | XP_011540925.1 | 2, 3 | 441 | |||
GenBank mRNA | AF533708.1 | - | 2, 3 | 433 | ||
AK125519.1 | - | 1, 2 | 256 | |||
AK313534.1 | - | 2, 3 | 336 | |||
AY358616.1 | - | 1, 2 | 543 | |||
BC036526.2 | - | 2, 3 | 428 | 5266665 | ||
DN999478.1 | - | 2, 3 | 482 |