Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RARB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000965.4 | NP_000956.2 | 2, 3 | 774 | ||
NM_001290216.2 | NP_001277145.1 | 5, 6 | 817 | |||
NM_001290217.1 | NP_001277146.1 | 2, 3 | 232 | |||
NM_001290266.1 | NP_001277195.1 | 2, 3 | 417 | |||
NM_001290276.1 | NP_001277205.1 | 2, 3 | 774 | |||
NM_001290277.1 | NP_001277206.1 | 2, 3 | 774 | |||
NM_001290300.1 | NP_001277229.1 | 2, 3 | 238 | |||
NM_016152.3 | NP_057236.1 | 2, 3 | 417 | |||
NR_110892.1 | 2, 3 | 774 | ||||
NR_110893.1 | 2, 3 | 774 | ||||
GenBank mRNA | AB307688.1 | - | 2, 3 | 305 | ||
AF157483.1 | - | 2, 3 | 416 | |||
AK292032.1 | - | 2, 3 | 324 | |||
AK299756.1 | - | 2, 3 | 392 | |||
AY501390.1 | - | 2, 3 | 206 | |||
BC060794.1 | - | 2, 3 | 456 | 30341884 | ||
CD014020.1 | - | 2, 3 | 321 | |||
CD014021.1 | - | 2, 3 | 321 | |||
CD014022.1 | - | 2, 3 | 321 | |||
DA882771.1 | - | 2, 3 | 232 | |||
DC376623.1 | - | 2, 3 | 392 | |||
DQ083391.1 | - | 2, 3 | 238 | |||
HQ692864.1 | - | 2, 3 | 305 | |||
X07282.1 | - | 2, 3 | 648 | |||
Y00291.1 | - | 2, 3 | 626 |