Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153809.2 | NP_722516.1 | 1 | 176 | ||
GenBank mRNA | AF390562.1 | - | 1 | 181 |