Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC38A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172702.1 | NP_001166173.1 | 2 | 302 | ||
NM_153811.2 | NP_722518.2 | 2 | 302 | |||
NR_033344.1 | 2 | 478 | ||||
XM_006720049.3 | XP_006720112.1 | 2 | 254 | |||
XM_011536469.2 | XP_011534771.1 | 2 | 254 | |||
XM_017021020.1 | XP_016876509.1 | 2 | 254 | |||
XM_017021021.1 | XP_016876510.1 | 2 | 254 | |||
XM_017021022.1 | XP_016876511.1 | 2 | 567 | |||
XM_017021023.1 | XP_016876512.1 | 2 | 254 | |||
XM_017021024.1 | XP_016876513.1 | 2 | 254 | |||
XM_017021025.1 | XP_016876514.1 | 2 | 545 | |||
XM_017021026.1 | XP_016876515.1 | 2 | 559 | |||
GenBank mRNA | AF070578.1 | - | 2 | 284 | ||
AF543422.1 | - | 2 | 254 | |||
AK303839.1 | - | 2 | 281 | |||
BC110378.1 | - | 2 | 163 | 5737780 | ||
BP352279.1 | - | 2 | 179 | |||
BX248072.1 | - | 2 | 123 | |||
BX648332.1 | - | 2 | 176 | |||
DC394244.1 | - | 2 | 478 | |||
DR002669.1 | - | 2 | 309 |