Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP2A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001681.3 | NP_001672.1 | 1 | 655 | ||
NM_170665.3 | NP_733765.1 | 1 | 655 | |||
XM_005253888.2 | XP_005253945.1 | 1 | 650 | |||
XM_011538402.2 | XP_011536704.1 | 1 | 650 | |||
GenBank mRNA | BC035588.1 | - | 1 | 202 | 5503508 | |
BI553880.1 | - | 1 | 555 | |||
KU177929.1 | - | 1 | 92 | |||
KU177930.1 | - | 1 | 92 | |||
KU177931.1 | - | 1 | 92 | |||
M23114.1 | - | 1 | 255 | |||
M23115.1 | - | 1 | 95 |